Imprimer
Membrane Microsart® e.motion
Fournisseur: Sartorius
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
11406Z-47-SCM 13006Z-47-SCM 13806Z-47-SCM 13906Z-47-SCM 13005Z-47-SCM 11406Z-50-SCM 114H6Z-47-SCM 13004Z-47-SCM 13005Z-50-SCM 13006Z-50-SCM 13806Z-50-SCM 13906Z-50-SCM 139H6Z-47-SCM 11407Z-47-SCM 11403Z-47-SCM 11407Z-50-SCM 15407Z-47-SCM 114H6Z-47-SFM 114H6Z-50-SCM 130H6Z-50-SCM 130H6Z-47-SCM 114H6Z-47-SJR 130H6Z-47-SJR
514-0076EA 405 EUR
514-0076 514-0077 514-0078 514-0079 514-0058 514-7018 514-7015 514-7021 514-7023 514-7024 514-7025 514-7026 514-0057 514-7019 514-7016 514-7020 515-0241 SART114H6Z-47-SFM SART114H6Z-50-SCM SART130H6Z-50-SCM 515-0231 SART114H6Z-47-SJR SART130H6Z-47-SJR
Membrane Microsart® e.motion
Filtres Filtres à membrane
These sterile cellulose nitrate and PES Microsart® e.motion membrane filters can be used for colony counting, particle testing and microscopy. They are specifically designed for use in the Microsart® e.motion dispenser.
  • Outstanding recovery rates for microorganisms
  • Product data is printed on individual envelopes
  • Gamma irradiated, 25 kGray
The special pleating of the band of membrane filters ensures that they are perfectly flat when dispensed. The membrane filters can also be used with nutrient pad sets.
Certifications: 0,45 µm filters are according to ISO 7704
Cdt: Individually sterile sealed in a format packaged on a pleated band, without protective paper.