Imprimer
Membrane filters, PTFE, WTP range, Whatman™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7582-002 7582-004 7585-004 7590-002 7590-004
514-0775EA 461 EUR
514-0775 516-1741 516-1742 514-0776 516-1743
Membrane filters, PTFE, WTP range, Whatman™
Filtres Filtres à membrane
Whatman PTFE membranes from Cytiva are strong hydrophobic membrane filters resistant to the corrosive solvents and aggressive media used in gas sterilization and liquid clarification.
  • Polypropylene grid support material for increased strength
  • Temperature resistant up to 120 °C
  • Resistant to most aggressive acids, alkalis, and solvents
  • Hydrophobic membrane filters can collect aqueous aerosols
PTFE membranes are especially effective for use with aggressive solvents, liquids, and gases.
PTFE membranes are used for liquid clarification of corrosives, solvents, and aggressive fluids. Effective sample filtration is critical to prevent equipment damage. Cytiva's PTFE hydrophobic membrane filters are also suitable for air and gas sterilization because of their ability to capture aqueous aerosols.