Imprimer
Capsules d'échantillonnage, Envirochek® et Envirochek® HV
Fournisseur: Cytiva (Formerly Pall Lab)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
12098 12099 12107
514-4087EA 5830 EUR
514-4087 514-4088 514-4089
Capsules d'échantillonnage, Envirochek® et Envirochek® HV
Filtres Capsules de filtration
For the concentration and recovery of Cryptosporidium oocysts and Giardia cysts from source or finished water. HV (high volume) capsule is designed for sampling up to 1000 litres of treated water and is validated for up to 50 litres of source water.
Envirochek®: PES hydrophile, Envirochek® HV: polyester hydrophile; boîtier: PC.
HV (high volume) capsule is designed for sampling up to 1000 litres of treated water and is validated for up to 50 litres of source water.