Imprimer
Synthetic filter, SUSP 70
Fournisseur: Ahlstrom-Munksjö
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
460001 460002 460003
512-6151EA 154 EUR
512-6151 512-6150 512-6149
Synthetic filter, SUSP 70
Filtres Filtres de fibres de verre
SUSP 70 is a woven polyamide fabric with a 70 μm mesh size used to capture coarse particles such as may be found in waste water.