Imprimer
Dispositifs de filtration sous vide multiple, Whatman™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
10444835 10445835 10498761 10498762
514-0218EA 8000 EUR
514-0218 516-0501 516-0418 516-0419
Dispositifs de filtration sous vide multiple, Whatman™
Collecteurs
Multiple vacuum filtration apparatus suitable only for vacuum operation.
  • For microbiological quality control
  • Residue analyses
  • Serial filtration carried out rapidly and easily with a common drainage outlet
Stainless steel including sieve filter support.
Individual stopcocks, 47/50 mm filter size, tubing nozzle 9 mm vacuum connection.
Attention: Suitable only for vacuum. Do not exceed 1.3bar (300mbar overpressure) when using flushing tubes.