Imprimer
Capsules filtrantes en fibres creuses MediaKap®
Fournisseur: Spectrum Laboratories
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ME2M02018S ME2M05018S ME2M05B12S ME2M10B12S ME2M25B06S ME2M50B03S
734-0868EA 1220 EUR
734-0868 734-0870 734-0871 734-0873 734-0874 734-0875
Capsules filtrantes en fibres creuses MediaKap®
Filtres Unités de filtration sous vide
MediaKap filters are designed to sterilise or clarify culture media or any other aqueous solutions. Can be used as pressure filters with a peristaltic pump, a hand pump or a syringe.
  • 0,2 µm, naturally hydrophilic
  • Non-pyrogenic
Sterile membrane made from hollow, mixed cellulose ester fibres
They reduce filtering times considerably (for example, 5 l of a serum-free culture medium can be filtered in less than 15 minutes using a MediaKap® 5).