Imprimer
Face shields, Bionic™
Fournisseur: HONEYWELL SÉCURITÉ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1011624 1011623 1011933
111-0134EA 101 EUR
111-0134 111-0133 111-0252
Face shields, Bionic™
Face Shields
This innovative face shield provides protection from extreme mechanical hazards. It can be worn in combination with safety spectacles or respiratory protection.
  • With extended protection for brow and chin
  • Locking mechanism for added safety
  • Extra soft padding with washable or replaceable sweatband
  • Various adjustments for best comfort
Weight: 355 g
EN 166, EN 169, EN 170, 1 F, B, T, C, 3, 8, 9
Certifications: EN 166, EN 169, EN 170, 1 F, B, T, C, 3, 8, 9