Imprimer
Disposable earplugs, Max®
Fournisseur: HONEYWELL SÉCURITÉ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3301130 3301161 1013046
111-0139EA 48.45 EUR
111-0139 111-0140 111-0394
Disposable earplugs, Max®
Earplugs Disposable Earplugs
Max® earplugs offer superior noise blocking performance in a disposable earplug.
  • Bell shape provides maximum ear comfort
  • Improved design is easier to insert
  • Smooth, soil resistant covering prevents dirt penetrating the surface prior to insertion
SNR: 37 dB
SNR: 37 dB
EN 352-2:2002
Certifications: EN 352-2:2002