Imprimer
Bottle top tilt measures
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3.580.301 3.580.302 3.580.310 3.580.360 3.580.315 3.580.320 3.580.325 3.580.330 3.580.340 3.580.305 3.580.350
613-8711EA 47.3 EUR
613-8711 613-8712 613-8714 613-8721 613-8715 613-8716 613-8717 613-8718 613-8719 613-8713 613-8720
Bottle top tilt measures
Bouchons mesureurs inclinés
Glass, with normal-shaped core.
  • For repetitive dispensing of the same volume
  • Can bleed with all bottles with NS 29/32