Imprimer
Instrument trays with lid
Fournisseur: BOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8400 8404 8406 8407
216-0016EA 79.2 EUR
216-0016 216-0017 216-0018 216-0019
Instrument trays with lid
Trays/Dishes/Pots Instrument Trays/Dishes
Stainless steel, corrosion-resistant, 18/10.
  • Special corner shape for fast, easy cleaning
  • Polished electrolytically, anti-magnetic
  • Good chemical resistance, very good thermal resistance
With a cover that enables the instrument dishes to be stacked.