Imprimer
Instrument trays
Fournisseur: GLASWARENFABRIK KARL HECHT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
42910010 42910003
HECH42910010EA 188 EUR
HECH42910010 HECH42910003
Instrument trays
Trays/Dishes/Pots Instrument Trays/Dishes
Glass with chrome-plated knobbed lid.
  • Can be sterilised in hot air up to 180 °C
  • Robust
  • Good chemical resistance
Multipurpose dishes with vertical walls for a variety of applications in the laboratory, field use and industry.