Imprimer
VWR®, Creusets, acier inoxydable 304
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
459-4501EA 37.9 EUR
459-4501 459-4502 459-4503
VWR®, Creusets, acier inoxydable 304
Creusets
These crucibles are made from stainless steel 304.
  • Highly resistant to chemicals and reagents
  • Withstand maximum temperature of 800 °C
Without lid.