Imprimer
Ice packs
Fournisseur: HONIGBERG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
418 420
216-7010EA 7.6 EUR
216-7010 216-7011
Ice packs
Cooling Packs
Heavy, metal-coated PE film filled with special gel cooling pad
  • Permanently leak-proof and puncture resistant
  • Content after thawing gel
  • Reusable, non-toxic
These durable and convient ice packs are designed for many applications like cooling, shipping and cold storage.
Note: Those -23 °C formulation gel packs can be placed in an insulated container at temperatures between 21 and 23 °C to keep the materials at or below 0 °C for over 40 hours.