Imprimer
Pots stériles, Falcon®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
354013 354015 354014 354020 354017
391-0020EA 140 EUR
391-0020 391-0023 392-0298 392-0396 392-0299
Pots stériles, Falcon®
Récipients de collecte d'échantillon Conteneurs à usage universel
PP, sterile, with or without PE lid.
  • Moulded-in graduations for easy measurement
  • Inert and chemically resistant to commonly used laboratory reagents at room temperature
Convenient, single-use containers for collection, transport and storage of a wide variety of specimens.
Polyethylene lid is frosted for easy marking, and fits both container sizes.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904