Imprimer
Condensers, Graham type, with Quickfit® ground joints
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C3/12/SC C3/14/SC C3/13/SC C3/22/SC C3/23/SC C3/24/SC
201-8280EA 121 EUR
201-8280 201-3041 201-8281 201-8282 201-8283 201-8284
Condensers, Graham type, with Quickfit® ground joints
Condenseurs
PYREX®, borosilicate glass.
  • Highly efficient condensers with a large cooling surface
  • High chemical and temperature resistance
Coil type, Graham pattern, suitable for either reflux or distillation work. Detachable plastic screw thread connector for the safe and easy attachment of flexible tubing.