Imprimer
Condensers, Inland Revenue, with Quickfit® ground joints
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C6/13/SC C6/12/SC
201-8293EA 396 EUR
201-8293 BIBBC6/12/SC
Condensers, Inland Revenue, with Quickfit® ground joints
Condenseurs
PYREX®, borosilicate glass.
  • Vapour and distillate move down through the coil
  • Coolant water flows outside the coil giving high cooling efficiency
  • High chemical and temperature resistance
Complete with detachable plastic hose connectors for the easy and safe attachment of flexible tubing.
Approximate surface area is 2,5 x 102 m2
Note: Inland Revenue condensers should be used in the vertical mode only with vapour fed in via the upper socket and liquid distillate leaving via the lower cone. Distillation rates should be slow to avoid "slugging" and back pressure in the coil.