Imprimer
Liebig condenser with standard ground joints
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
05210201 05210202 05210301 05210302 05210303 05210304 05210402 05210403 05210404 05210504 05210401 05210704
201-0352EA 64.2 EUR
201-0352 201-0353 201-3313 201-0354 201-0355 201-3312 201-0356 201-0357 201-0358 201-0359 LENZ05210401 LENZ05210704
Liebig condenser with standard ground joints
Condensers
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours, standard ground joint cone and sleeve. Water connection via glass olive.
Certifications: DIN 12576