Imprimer
Air condensers with Quickfit® ground joints
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C2/22 C2/11 C2/12
201-8279EA 62 EUR
201-8279 BIBBC2/11 BIBBC2/12
Air condensers with Quickfit® ground joints
Condenseurs
PYREX®, borosilicate glass.
  • High chemical and temperature resistance
  • Both cone and sockets are the same size
For condensation of materials with boiling points above 150 °C.
Certifications: ISO 1773, BS 6352