Imprimer
Avantor® Hichrom, Connecteurs
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
554-7715EA 36.4 EUR
554-7715 554-7714
Avantor® Hichrom, Connecteurs
Raccords et connecteurs
The HI-065 female-female connector consists of a 60 mm length of 1/16" (0,007 mm) PEEK tubing with PEEK fingertight fittings and is suitable for connecting to all end fittings.
The HI-061 male-male connector provides an ultra low dead volume and comes complete with two PEEK end fittings.
  • Low dead volume
  • Male-male and female-female options
  • Inert and biocompatible