Imprimer
Ferrules type OGF
Fournisseur: Trajan Scientific and Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0726003 072604 072605 072606 072607 072608 072609 072610 072613 072615 0726151 072617 072621 0726212 072622 072623 072625 072626 072627 0726270 072628 0726281 0726282 0726283 0726520
SGEA0726003EA 58.2 EUR
SGEA0726003 SGEA072604 SGEA072605 SGEA072606 562-1073 SGEA072608 SGEA072609 562-1076 SGEA072613 SGEA072615 SGEA0726151 SGEA072617 558-0176 SGEA0726212 558-0178 558-0179 558-0180 558-0181 558-0182 558-0183 558-0184 558-0185 558-0186 558-0187 558-0193
Ferrules type OGF
Raccords et connecteurs
These reusable ferrules are made from 100% graphite. They have a temperature limit of 450 °C.
  • Easy to use, forms a reliable seal
  • Soft material, forms a soft grip to the capillary column
  • Low emissions
Poorly defined or selected connections can lead to an increase in dead volume, leaks from cycled temperatures and mismatched tubing sizes. Selecting the correct GC connections will improve your chromatography.