Imprimer
Combinaisons résistantes aux produits chimiques, Spacel 2000 EBJ, PE
Fournisseur: HONEYWELL SÉCURITÉ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4180130/2 4180130/3 4180130/4 4180130/6
113-1620EA 0 EUR
113-1620 113-1621 113-1622 113-1624
Combinaisons résistantes aux produits chimiques, Spacel 2000 EBJ, PE
Vêtements à usage unique
This overall made from PE features hood and zip and offers protection from low risks in a work environment, e.g. from harmful substances whose effects can be eliminated (cleaning products which are not too aggressive, diluted detergents etc.) or poor weather or for industrial cleaning operations.
  • Elasticated at wrists and ankles, for especially secure hold at these points
  • Perfect fit thanks to elasticated back
  • 'Spaseal' sealing, no puncturing, thus offering improved protection
Thickness: 50 µm
Colour: blue
Certifications: EN 340, cat. I