Imprimer
Tubing clamps
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F182270000 F182280000 F182290000
229-0243EA 111 EUR
229-0243 229-0320 229-0321
Tubing clamps
Clamps Tubing and Sanitary Clamps
Plastic. These tubing clamps are durable and will not rust or corrode. Three sizes are available for best tubing fit.
  • Mini tube clamp with single position on/off gives positive shut-off with fingertip control; use for thin-walled tubing under 4,7 mm (³⁄₁₆") Ext.Ø
  • Mid-range clamp has a 15-position stepped adjustment that controls flow from full open to complete closure; fits tubing from 3,2 to 11 mm (⅛ to ⁷⁄₁₆") Ext.Ø
  • Maxi clamp has a 12-position ratchet control from full open to complete closure; fits tubing up to 19 mm (¾") Ext.Ø
  • Autoclavable