Imprimer
Clips for conical standard ground joints
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
01103514 01103529 01103519 01103524 01103545 01103599
201-1079EA 25.1 EUR
201-1079 201-1082 201-1080 201-1081 201-1083 201-1084
Clips for conical standard ground joints
Clamps Standard Taper and Spherical Joint Clamps
Nickel-chromium steel. For conical ground joint connections.
  • Quick, easy assembly and disassembly
  • Very good thermal resistance, good chemical resistance
Informations de commande: Optional fixing screws are available.