Imprimer
Burette clamps
Fournisseur: USBECK Laborgeräte
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2861 2862
241-7461EA 35.5 EUR
241-7461 241-7462
Burette clamps
Clamps General Purpose Clamps
Die-cast aluminium. For simple and secure fixing of burettes.
  • With bosshead for fixing to retort stand
  • Span of clamp 0 to 20 mm
  • With roll holders