Imprimer
Inoculating loops, Microstreaker
Fournisseur: MEDICAL WIRE EQUIPMENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MW180 MW182 MW184
300-0501EA 60.4 EUR
300-0501 391-0471 391-0472
Inoculating loops, Microstreaker
Étaleurs Aiguilles et inoculateurs
Inoculation loop of nickel-chromium wire of not more than 60 mm length, conforming to Howie recommendations. Colour coded handle.