Imprimer
Automatic burettes, Pellet pattern, without intermediate stopcock, Schellbach, BLAUBRAND®
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
22521 22522 22523
612-5402EA 268 EUR
612-5402 612-5403 612-5404
Automatic burettes, Pellet pattern, without intermediate stopcock, Schellbach, BLAUBRAND®
Burettes
Borosilicate glass 3.3, class AS, DE-M marking.
  • Calibrated to deliver 'Ex'
  • Schellbach stripe, ring marks at major graduations
  • Automatic zeroing, needle-valve stopcock with PTFE spindle without intermediate stopcock
DIN EN ISO 385
Certifications: DIN EN ISO 385
Informations de commande: Supplied with a batch certificate. Burettes are also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.