Imprimer
Automatic burettes, Dr. Schilling pattern, SILBERBRAND
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
23753 23755 23756 23758 23766 23768
612-9920EA 141 EUR
612-9920 612-9921 612-9922 612-9923 BRND23766 612-5405
Automatic burettes, Dr. Schilling pattern, SILBERBRAND
Burettes
AR-GLAS®, class B.
  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe, reduced distance between subdivision marks
  • Automatic zeroing, squeezing bottle allows rapid filling, micrometre screw allows fine titration
With bottle: 500 ml for 10 and 15 ml; 1000 ml for 25 and 50 ml.
Avec flacon: 500 ml pour burette 10 et 15 ml; 1000 ml pour burette 25 et 50 ml
Certifications: Error limits according to class B, DIN EN ISO 385.