Imprimer
Ice buckets with lids, Azlon®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
IBB001P IBB002P IBB003P
216-6999EA 141 EUR
216-6999 216-2822 216-2823
Ice buckets with lids, Azlon®
Ice Containers
Polyurethane.
  • Three colours available for better separation of work areas by colour
  • Octagonal shape that is stackable and stable with integral handles
  • Unbreakable in normal use
Excellent insulating properties suitable for use with dry ice, water ice and salt solutions.
Capacity: 4,5 Litres
Weight: 1,2 kg