Imprimer
Storage box, with snap-on lid, Azlon®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BWY032 BWY034 BWY038 BWY039 BWY030 BWY036
216-2814EA 20 EUR
216-2814 215-0396 215-0398 216-2816 216-2813 215-0397
Storage box, with snap-on lid, Azlon®
Boîtes Boîtes de rangement
HDPE, translucent.
Suitable for food contact applications in accordance with EU regulations.
  • Lightweight and unbreakable in normal use
  • Tight fitting snap on lids create an airtight seal
  • Suitable for general and refrigerated storage down to −50°C