Imprimer
PCR-Cooler
Fournisseur: EPPENDORF
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3881000.015 3881000.023 3881000.031 0030124.545
479-1160EA 121 EUR
479-1160 479-1161 479-1162 713-0004
PCR-Cooler
Coolers Benchtop Coolers
Cools sensitive samples for more than one hour at 0 °C.
  • In universal 96-well format for 0,2 ml PCR-tube strips, plates and single tubes
  • A colour change from violet to pink or dark blue to light blue shows when the temperature has exceeded 7 °C
Coolers for cool samples throughout PCR setup and racks for safe sample transport and storage.