Imprimer
Bottles, wide neck, with screw cap
Fournisseur: MURRAY S
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
P201/26/P515/3 P201/24P500/14 P201/25P515/39 P201/22P500/13
215-0973EA 146 EUR
215-0973 215-3368 215-3371 215-3367
Bottles, wide neck, with screw cap
Flacons Flacons en plastique
HDPE, white with black PP closure.
  • Good chemical resistance, good temperature resistance
  • Extra wide opening for easy filling and emptying
  • Low profile, lightweight
Ideal for liquids, pastes, and powders.