Imprimer
Flacons carrés à col large et fileté
Fournisseur: Kautex
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
310-74374 310-74372 310-74373 310-74375 310-84404 310-74371
216-3907EA 3.4 EUR
216-3907 215-3120 216-3900 216-3904 216-3906 215-2287
Flacons carrés à col large et fileté
Récipients de collecte d'échantillon Conteneurs à usage universel
PETG. Excellent transparency. Black closure with PE-foam wad.
  • Wide neck opening
  • Easy to fill, easy to clean
  • Leakproof
  • Suitable for food
  • Crystal-clear for an excellent product presentation
  • Halal certified
This container is also suitable as a sales or gift packaging.