Imprimer
Flacons, col large, avec bouchon à vis, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2120-0005 2120-0010 2121-0005 2120-0005Z 2120-0010Z 2121-0010
215-7351EA 28.9 EUR
215-7351 215-7352 215-7361 NALG2120-0005Z NALG2120-0010Z 215-1281
Flacons, col large, avec bouchon à vis, Nalgene®
Flacons Flacons en plastique
En PE-HD translucide, bouchon en PP/En PPCO translucide, bouchon en PP.
  • HDPE: Good chemical resistance, heat resistant up to 120 °C
  • PPCO: Very good chemical resistance, heat resistant up to 121 °C, autoclavable
  • Extra wide opening
Ces flacons universels grand volume permettent de nombreuses applications. Etanchéité non garantie.
These high-density, polyethylene bottles are good for use in the freezer to −100 °C (−148 °F). Chemical-resistant HDPE is compatible with a wide variety of lab reagents.
Linerless cap closes the bottle without a liner that can wrinkle, cause leaks and contaminate your solutions. This bottle is not liquid leakproof. Bottle and carton are readily recyclable in most communities as they are manufactured from non-toxic materials.
Informations de livraison: Includes 100 mm polypropylene screw caps.