Imprimer
Bottles, wide mouth, rectangular, with screw cap, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2007-0004 2007-0008 2007-0016 2007-0032 2007-0064 2009-0004 2009-0008 2009-0016 2009-0032 2009-0064
215-7313EA 39 EUR
215-7313 215-7314 215-7315 215-7316 215-7317 215-7323 215-7324 215-7325 215-7326 215-7327
Bottles, wide mouth, rectangular, with screw cap, Nalgene®
Bottles Plastic Bottles
HDPE, translucent or HDPE, amber with PP screw cap.
  • Good chemical resistance
  • Heat resistant up to 120 °C
  • Leakproof
Ideal for collecting, storing and shipping samples or storing liquids and dry substances. Space saving rectangular base. Amber bottles protect light-sensitive substances.