Imprimer
Wash bottles, wide neck, Unitary ™, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DS2408-0250 DS2408-0500
215-8131EA 58.1 EUR
215-8131 215-8132
Wash bottles, wide neck, Unitary ™, Nalgene®
Bottles Wash Bottles
Bottle and riser made from red LDPE, with red PP screw cap.
  • Wide neck for quick, safe filling, non-drip
  • Formed from a single piece, riser can no longer become contaminated or leak
  • Good chemical resistance, heat resistant up to +80 °C