Imprimer
Pissette intégrale à col étroit, Azlon®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BGF130P BGF136P
215-0490EA 32.9 EUR
215-0490 215-0494
Pissette intégrale à col étroit, Azlon®
Flacons Pissettes
LDPE, translucent, with PP spray cap.
  • Slim hexagonal cross section makes the bottles particularly suited to smaller hands
  • Precision tip is ideal for use as a dropper or for fine liquid jet dispensing
Ergonomic wash bottles with an integral moulded dispensing tube and removable jet tip.