Imprimer
Safety wash bottles, wide neck, with printed name
Fournisseur: Kautex
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
303-770018 303-770021 303-770022 303-770023 303-770024 303-770025
215-3333EA 6.7 EUR
215-3333 215-3336 215-3337 215-3338 215-3339 215-3335
Safety wash bottles, wide neck, with printed name
Flacons Pissettes
LDPE, Round, Natural colour.
  • Printed-on hazard symbols
  • Abrasion-resistant printing
  • Sliding delivery tube
Substances of content in German, English, French.