Imprimer
Flacons ronds avec bouchon à filetage, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
218011453 218011753 218012458 218013651 218014459 218015455 218017353 218018658 218019157 218018855 218013651Z 218014459Z 218015455Z 218016357Z 218012458Z 218012955 218015155 218016957 218010851 218016357 218019251
215-1512EA 21.5 EUR
215-1512 215-1513 215-1514 215-1515 215-1516 215-1517 215-1519 215-1520 215-1522 215-1521 SCOT218013651Z SCOT218014459Z SCOT218015455Z SCOT218016357Z SCOT218012458Z 215-2428 215-2429 215-2430 215-3214 215-1518 215-4775
Flacons ronds avec bouchon à filetage, DURAN®
Flacons Flacons en verre
En verre borosilicaté 3.3 transparent DURAN®, bouchon à visser en PP.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
Round, with graduation, DIN thread, pouring ring and blue screw cap.
With printed Retrace Code (batch identification).
*With specially shaped glass rim for improved pouring; no additional pouring ring is needed.
**Ring-shaped reinforcement on the bottle shoulder makes the filling line of theEn savoir plus