Imprimer
Bottles, narrow mouth, with screw cap, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2006-0002 2006-0004 2006-0016 2006-0032 2006-9025 2006-9050 2006-9125 2006-0001 2006-0008
215-7565EA 21.8 EUR
215-7565 215-7566 215-7570 215-7571 215-7562 215-7563 215-7561 215-7564 215-7568
Bottles, narrow mouth, with screw cap, Nalgene®
Bottles Plastic Bottles
PPCO, translucent with PP screw cap.
  • Excellent chemical resistance
  • Heat resistant up to +121 °C, autoclavable with or without contents
  • Robust, leakproof
Ideal for storing and shipping liquids.
Attention: *The product(s) on this page are outside the scope of the EU Directive 2019/904.