Imprimer
Bottles, narrow mouth, with screw cap, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2015-0030 2015-0060 2015-0125 2015-1000 2015-0250 2015-0500 2015-2000
215-7601EA 50.7 EUR
215-7601 215-7602 215-7603 215-7606 215-7604 215-7605 215-7607
Bottles, narrow mouth, with screw cap, Nalgene®
Bottles Plastic Bottles
PC, transparent with PP screw cap.
  • Robust, impact resistant, light
  • Heat resistant up to +135 °C, autoclavable
  • Leakproof
Ideal for storing aqueous solutions and for short-term storage of culture media. The 2000 ml size includes an integral handle for easier handling.
Made only from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. Excellent clarity helps easily identify content levels.
Molded in graduations for ease in dispensing. 2 L size has molded-in handgrips to aid in lifting and pouring contents.
Certifications: USP Class VI; FDA - 21 CFR