Imprimer
Whatman™ Cellulose Blotting Papers, Grade GB003, Whatman products (Cytiva)
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
10426880 10426890 10426892 10427818 10427826 10427810 10427812 10427813 10427804 10427806 10547922 10427805
732-4091EA 54.6 EUR
732-4091 732-4092 732-4093 732-4098 732-4100 732-2531 732-4229 732-4230 732-2760 732-2599 732-2762 732-2761
Whatman™ Cellulose Blotting Papers, Grade GB003, Whatman products (Cytiva)
Papiers buvard
This general purpose blotting paper is made from pure raw materials with a high absorbency and is used as a membrane gel support. Due to its thickness (0,8 mm), it is recommended for the lysis/denaturation of colony or plaque lifts and Western blots.
  • Pure cellulose produced entirely from the highest quality cotton linters with no additives ensures that no contamination will occur during the transfer steps
  • Manufactured and tested specifically for chromatographic and blotting techniques ensuring the wicking capability and uniformity of capillary action that are important in obtaining clean and even transfers during blotting
  • Industry standard for blotting procedures
  • Convenient sizes available in sheets precisely cut to the most popular gel and transfer membrane sizes allows 'out-of-the-box' usage and eliminates sheet-to-sheet variation
A thick paper recommended for the lysis/denaturation of colony or plaque lifts and Western blots.