Imprimer
Beakers, low form, Azlon®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BDD312P BDD318P BDD324P BDD330P BDD336P BDD340P
213-0452EA 71.8 EUR
213-0452 213-0451 213-0456 213-0455 213-0453 213-0449
Beakers, low form, Azlon®
Béchers
PMP (TPX), crystal clear.
  • Excellent chemical resistance
  • Heat resistant up to +180 °C
  • Smooth surfaces for quick, easy cleaning
Ideal for general laboratory use. With spout and exceptionally easy to read red graduation. Straight sided.
ISO 7056
Certifications: ISO 7056