Imprimer
Autoclave bags
Fournisseur: Greiner Bio-One
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
643201_500 646201_500 649201_350 645201_500 BAG1_200 BAG2_200 BAG3_200
129-1750EA 344 EUR
129-1750 129-1752 129-1753 GREI645201_500 129-1754 129-1755 129-1756
Autoclave bags
Bags Waste Disposal Bags
These disposal bags are used for storing contaminated petri dishes or test tubes.
  • Suitable for autoclaving by wet steam
  • Specially designed to eliminate contaminated waste in the laboratory
PP, transparent, thickness: 50 µm