Imprimer
Aluminium foil
Fournisseur: Alujet-Universal
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
52031531 52031545 52092045 52151045 52151050 52151055 52151060 52185050 520301530 52151025 521501030 52151035 52151040
293-4175EA 91.1 EUR
293-4175 293-4176 293-4177 293-4178 293-4179 293-4180 293-4181 293-4182 293-4183 293-4184 293-4185 293-4186 293-4187
Aluminium foil
Aluminium Foils
Pure aluminium 99,0 to 99,5%. Ideal for packaging, covering and insulation.
  • Economical large-size rolls for use in a roll dispenser
  • Available in various widths and thicknesses
  • Length of roll to suit various requirements