Imprimer
Vaporisateur d'air comprimé
Fournisseur: HM INDUSTRIESERVICE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
100093 100545
291-1201EA 47.2 EUR
291-1201 HMIN100545
Vaporisateur d'air comprimé
Aérosols
For the quick and easy removal of dust and small amounts of dirt in hard to reach places. For use in medical engineering, metrology, microscopy, precision engineering and in the fields of photography and optics.
  • Includes slip-on capillary tubes (15 cm long) for highly precise spraying
  • Powerful, dry jet of air that is free from oil
  • Non flammable, does not contain any CFCs