Imprimer
Multiple adapters
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
05010011 05010022 05010033 05010044
201-2215EA 39.6 EUR
201-2215 201-2218 201-0337 201-2216
Multiple adapters
Distillation Components Multiple Distillation Adapters
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Two sockets and one cone, standard ground joints and two parallel necks.