Imprimer
Flange clamps
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
06248515 06248525 06248550
201-0120EA 103 EUR
201-0120 LENZ06248525 LENZ06248550
Flange clamps
Pinces Pinces à usage général
Plastic, black.
  • Easy to assemble and disassemble
  • Tough construction, good temperature resistance
  • Matching gaskets are available as an option
For fast, secure connection of ball or flat flanges. No gasket.