Imprimer
Méthylcellulose, 4000 mPa.S (2% H₂O), blanc, en poudre
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0215549691 0215549680 0215549683 0215549690
ICNA0215549691EA 245 EUR
ICNA0215549691 ICNA0215549680 ICNA0215549683 ICNA0215549690
Méthylcellulose, 4000 mPa.S (2% H₂O), blanc, en poudre
Methyl cellulose
An aqueous solution is best prepared by dispersing the granules in hot (but not boiling) water with stirring and chilling to +5 °C. The solution is then stable at room temperature. Presence of inorganic salts increases the viscosity.
Prepared from wood pulp by treatment with alkali and methylation of the alkali cellulose with methyl chloride.
Methyl cellulose is a chemical compound derived from cellulose. It is a hydrophilic white powder in pure form and dissolves in cold (but not in hot) water, forming a clear viscous solution or gel. It is used as a thickener and emulsifier in various food and cosmetic products. It is not digestible, not toxic, and not an allergen.
1 mPa.s = 1cP
Température de stockage: Température ambiante
Numéro MDL: MFCD00081763
Numéro CAS: 9004-67-5