Imprimer
Cell culture dishes, Nunc™ UpCell™ surface
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
174902 174903 174904 174905 174906
734-2381EA 412 EUR
734-2381 734-2383 734-2253 734-2382 734-2384
Cell culture dishes, Nunc™ UpCell™ surface
Boîtes de culture Boîtes de Petri pour culture tissulaire
With lid, sterile. Single-use vented dishes feature a temperature-responsive cell culture surface that is ideal for culture passaging, single-cell analyses, and cell transplantation research.
  • Surface allows harvesting of cells with intact surface proteins
  • High cell viability is achieved with minimal hands-on time and without physical force
  • Cell surface proteins are preserved as no trypsinisation required
  • Releases adherent cells by reduction of temperature of the cell culture
  • Enables harvesting of cell sheets and creations of 3D tissue models held together by normal cell junctions and extracellular matrix