Imprimer
Frocks, PP
Fournisseur: Medicom
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
40.301/05 40.301/04 40.301V/04 40.301V/05
HOPE40.301/05EA 0 EUR
HOPE40.301/05 HOPE40.301/04 HOPE40.301V/04 HOPE40.301V/05
Frocks, PP
Vêtements à usage unique
These single-use hygiene frocks made from PP protect the working environment from particles coming from the operator's body (dander, hair, perspiration, etc.).
  • With collar and long sleeves
  • Elasticated cuffs
  • Snap buttons
  • Without pockets
Weight: 40 g/m²